Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPA™ Cobra™ FluoroSolv™ Gloves
Click to view available options
Size:
8
9
10
11
Length (English):
11 in.
14 in.
Length (Metric):
28 cm
35.5 cm
Description
Unsupported powder-free fluoroelastomer/nitrile gloves resist aromatic solvents, chloroform, and PCBs. Suitable for use in refineries, plating operations, laboratories, and hazardous material handling.
- Consist of a layer of black FluoroSolv fluoroelastomer polymer over a green, powder-free nitrile shell
- Two-tone layering serves as a visual wear indicator
- Nitrile layer also provides increased cut, puncture, and tear resistance
- Z-pattern texture channels chemicals away from glove surface, improving wet and dry grip
- Hand-specific and unlined with a flat cuff
- Embossed with size measurement for easy identification
- Style F-466 is 11 in. long (28 cm), 15 mil thick
- Style F-467 is 14 in. long (36 cm), 25 mil thick
Not for use with ketonic solvents.
Specifications
Powdered | Powder Free |
Size | 9 |
Length (English) | 11 in. |
Length (Metric) | 28 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction