MAPA™ Krytech Max 596 Cut-Resistant Gloves
For intricate handling of materials with dexterity
Manufacturer: MAPA™ 596128
 View more versions of this product
Catalog No. 19-249-811
  Check Availability
Add to Cart
    Specifications
    Cut Resistance Rating Level 5 in EN 388 and 38 N in ISO 13997
    Length (Metric) 9.5 to 11 in.
    Material Polyurethane
    Size 8
    Color Black / Purple
  • Description & Specifications
Related Products
Product Content Feedback