Learn More
MAPA™ NiProTect™ Controlled Environment Nitrile Gloves
Critically Clean 100% nitrile formulation.
Manufacturer:  MAPA™ 529428
 View more versions of this product
Catalog No. 19-050-338D
Add to cart
Description
Description
Meet the individual cleanliness level of your controlled environment. The 100% nitrile formulation eliminates protein allergy concerns while exhibiting excellent ESD properties and chemical splash protection. At 4mil thick, this glove delivers maximum tactility with good durability. Textured fingertips and unique finish enhance wet and dry gripping. Rolled cuff edge reduces glove roll-down and tearing when donning.
Available in critically clean (CC) for ISO Class 4 (formerly Class 10) compatibility. Ambidextrous.
Order Info
Sold in packs of 100.
Compliance
Meet or exceed ASTM D 3578 and ASTM D 130.
Specifications
Specifications
Safety
Textured
Ambidextrous
Large
White
30.48 cm
ISO Class 4
Rolled
Nitrile
4 mil
12 in.