MAPA™ PROtector™ Nitrile-Natural Rubber Gloves

Green natural rubber with a blue nitrile blend overdip in the work area

Manufacturer: MAPA™ 293429

 View more versions of this product

Catalog No. 11-388-115D

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Straight
    For Use With (Application) food handling and pharmaceutical applications
    Length (English) 13 in.
    Length (Metric) 33cm
    Material Nitrile, Natural Rubber
    Liner Material Cotton Flock
    Size X-Large
    Thickness 28mil
    Coating Material Embossed chained pattern
    Color Green, blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit