MAPA™ StanSolv™ Knit-Lined Nitrile Gloves
Durable, flexible gloves with insulating cotton lining
Manufacturer: MAPA™ 381410
 View more versions of this product
Catalog No. 11-392-6C
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR 170-199
    Glove Cuff Style Flared
    Hem Color Pink
    For Use With (Application) automotive, mechanical industry, surface treatment, part degreasing, automotive assembly, chemical industry, handling drums of chemical product, chemical product blending, manipulating valves
    Length (English) 14 in.
    Length (Metric) 35.5cm
    Lining Cotton knit
    Material Nitrile
    Palm Type Contoured
    Size 10
    Thickness 33mil
    Coating Material Embossed
    Color Green
  • Description & Specifications
Related Products
Product Content Feedback