Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ StanZoil™ ChemPly™ Neoprene Gloves

Ideal for acid handling, degreasing and many other applications

Manufacturer:  MAPA™ 413701

 View more versions of this product

Catalog No. 19-048-517H


Add to cart

Description

Description

Highly impervious to moisture, vapors and liquids, and feature outstanding resistance to numerous solvents, oils, acids, alcohols, alkalis, petroleum derivatives and animal fats.

  • Flared cuff design lets air circulate and ensures easy donning and doffing
  • “Z” pattern finish channels fluids from surface, provides excellent gripping
  • Multilayer, two-color (black over coral) construction prevents pinholes
  • Coral interior also serves as a wear indicator—shows when replacement gloves are needed
  • Curved finger design

Order Info

Styles N-360, N-440, and N-540 are 14 in. (36cm) long.
Styles N-730 and N-740 are 18 in. (46cm) long for protection up to the elbow for work in deep liquid applications.

Compliance

Comply with 21CFR170-199. 30mil and 40mil thick gloves are manufactured to meet NFPA standard for “Liquid Splash Protection Garments” for Hazardous Chemical Emergencies (Level B).

Specifications

Specifications

Safety
Flared
Unlined
11
Black
45.72 cm
Embossed Z-pattern
hazmat and chemical spills, Metal finishing, transformer rebuilding, agricultural chemicals, grinding/filing, transmission assembly, general plant maintenance, Petroleum refining and printing industry
Neoprene
22 mil
18 in.
21 CFR 170-199, NFPA 1992-1994
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit