Learn More
MAPA™ StanZoil™ NL-34/334 Chemical Resistant Polychloroprene Gloves
Chemical resistant gloves with enhanced rough grip and absorbent interlock liner are ideal for handling and transporting chemicals, as well as maintenance applications in wet environments.
$18.17 - $154.50
Specifications
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 407:X2XXXX
Material Neoprene
Color Black, Blue
Length (English) 12 in.
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-392-31A
View Documents
MAPA™
334946
6
Dozen of 12 for $81.12
Case of 6 DZ for $600.00
Add to cart
11-392-31B
View Documents
MAPA™
334947
7
Dozen of 12 for $154.50
Case of 6 DZ for $883.00
Add to cart
11-392-31C
View Documents
MAPA™
334948
8
Dozen of 12 for $18.17
Save 88%
Reg : $154.50
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Case of 6 DZ for $109.03
Save 88%
Reg : $883.00
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to cart
11-392-31D
View Documents
MAPA™
334949
9
Dozen of 12 for $154.50
Case of 6 DZ for $883.00
Add to cart
Description
Description
  • Soft, absorbent interlock knit cotton liner
  • Low-temperature protection
  • 1.5 AQL
  • Straight cuff, rough grip
  • Ideal for mechanical, chemical and petroleum industry applications
Specifications
Specifications
21 CFR 170-199
Neoprene
12 in.
Handling and transporting chemicals, maintenance in wet environments that include water, oil, greases and hydrocarbons
Lined
EN 407:X2XXXX
Black, Blue
30.5 cm
Rough
Interlock Cotton