Learn More
MAPA™ StanZoil™ Knit-Lined Multi-Layer Supported Neoprene Gloves
Combine flexibility with abrasion, cut and tear resistance. MAPA™ StanZoil™ Knit-Lined Multi-Layer Supported Neoprene Gloves have unique multi-dip construction offers liquid-proof hand protection and excellent chemical resistance to most oils, grease, acids, caustics, alcohols, and solvents.
Manufacturer:  MAPA™ 336429
 View more versions of this product
Catalog No. 19-039064
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Ideal for applications involving assembly, degreasing, lubricating, castings, steam cleaning, motor parts, chemical processing, petroleum refining, grinding, filing, metal finishing and general maintenance
  • Neoprene overdip in glove work area eliminates pinholes and weak spots throughout the glove and cuff
  • Nonslip finish ensures a safe grip—even on wet or oily parts
  • Two-way stretch cotton interlock knit lining insulates from heat and cold and absorbs perspiration
  • Seamless liner in work area protects hands from chafing
  • Roomy thumb crotch for long-wear comfort
  • Pinked cuff
  • Washable
Compliance
Meet 21CFR 170-199.
Description & Specifications
Specifications
Non-slip
chemical processing, Petroleum refining, grinding and filing of castings, Metal finishing, general maintenance
Neoprene
USDA, 21 CFR 170-199, CE
12 in.
Pinked
Interlock Cotton knit
10
Dark Green,Light Green
30.5cm