MAPA™ StanZoil™ Knit-Lined Multi-Layer Supported Neoprene Gloves

Combine flexibility with abrasion, cut and tear resistance. MAPA™ StanZoil™ Knit-Lined Multi-Layer Supported Neoprene Gloves have unique multi-dip construction offers liquid-proof hand protection and excellent chemical resistance to most oils, grease, acids, caustics, alcohols, and solvents.

Manufacturer: MAPA™ 334947

 View more versions of this product

Catalog No. 11-392-31B

  Check Availability
Add to Cart

    Specifications

    Certifications USDA, 21 CFR 170-199, CE
    Glove Cuff Style Pinked
    For Use With (Application) chemical processing, Petroleum refining, grinding and filing of castings, Metal finishing, general maintenance
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Lining Interlock Cotton knit
    Material Neoprene
    Size 7
    Coating Material Non-slip
    Color Black, blue

    • Ideal for applications involving assembly, degreasing, lubricating, castings, steam cleaning, motor parts, chemical processing, petroleum refining, grinding, filing, metal finishing and general maintenance
    • Neoprene overdip in glove work area eliminates pinholes and weak spots throughout the glove and cuff
    • Nonslip finish ensures a safe grip—even on wet or oily parts
    • Two-way stretch cotton interlock knit lining insulates from heat and cold and absorbs perspiration
    • Seamless liner in work area protects hands from chafing
    • Roomy thumb crotch for long-wear comfort
    • Pinked cuff
    • Washable

    Compliance

    Meet 21CFR 170-199.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit