Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Technic/NS-450 Neoprene/Natural Rubber Gloves

Manufacturer:  MAPA™ 450440

 View more versions of this product

Catalog No. 19-050-574


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Embossed Diamond
food processing, meat and poultry, chemical sampling, commercial housekeeping, inspection, Metal finishing, automotive painting, degreasing, plating, Bleaching, Leather tanning, commercial pot scrubbing, janitorial, maintenance
Latex, natural rubber,Neoprene
30mil
Non-sterile
40.64cm
Flat
Cotton Flock
10
Black
16 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit