MAPA™ Tri-Poly Powder-Free Trilite™ Grippy Gloves

New revolutionary outer finish provides extraordinary gripping power

Manufacturer: MAPA™ 983959

 View more versions of this product

Catalog No. 19-249-535

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex and is not recommended for use by those who may be allergic to natural rubber proteins


    Specifications

    Disposable Yes
    Exterior Finish Textured
    Glove Cuff Style Rolled
    For Use With (Application) laboratory, research facilities, janitorial, cleaning, small parts assembly, sorting applications
    Length (English) 11.5 in.
    Length (Metric) 29cm
    Material Tri-polymer
    Powdered No
    Size X-Large
    Thickness 6mil
    Color Purple

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit