MAPA™ Ultrane™ 550 Gloves
Combine comfort with sensitivity
Manufacturer: MAPA™ 550418
 View more versions of this product
Catalog No. 19-999-360
  Check Availability
Add to Cart
    Specifications
    Certifications CE, AQL 1.5, ASTM 3577
    Glove Cuff Style Knit
    For Use With (Application) excellent nonslip grip, abrasion resistance and smudge-free parts handling
    Laboratory/Cleanroom Use Yes
    Cleaning Procedure Launderable
    Glove Cuff Color Brown
    Length (English) 9.5 in.
    Length (Metric) 24cm
    Material Polyamide
    Size 8
    Coating Material Polyurethane Foam
  • Description & Specifications
Related Products
Product Content Feedback