Learn More
MAPA™ Ultrane™ 550 Gloves
Combine comfort with sensitivity
Manufacturer:  MAPA™ 550429
 View more versions of this product
Catalog No. 19-999-361
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyurethane Foam
Knit
Yes
9
Yellow
25.5cm
Launderable
excellent nonslip grip, abrasion resistance and smudge-free parts handling
Polyamide
CE, AQL 1.5, ASTM 3577
10 in.