MAPA™ Pylox™ Vinyl Bisque Grip Gloves
Combining chemical and mechanical protection with dexterity
Manufacturer: MAPA™ 541426
 View more versions of this product
Catalog No. 11-393-90
  Check Availability
Add to Cart
    Specifications
    Certifications CE
    For Use With (Application) intricate assembly, film processing, biological research, Resin application, Epoxy layup and furniture finishing, liquid tight gloves outperform natural Rubber and Nitrile in Acids, Caustics, Alcohols, Aldehydes, Oils and G
    Exterior Finish Bisque
    Glove Cuff Style Pinked
    Hand Left
    Length (English) 10.5 in.
    Length (Metric) 26.7cm
    Material Vinyl
    Powdered Yes
    Size Small
    Thickness 5mil
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback