MAPA™ Pylox™ Vinyl Gloves

Sulfur- and sodium-free

Manufacturer: MAPA™ 542426

 View more versions of this product

Catalog No. 11-392-37A

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Bisque
    Glove Cuff Style Pinked
    Hand Hand-specific
    For Use With (Application) intricate assembly, film processing, biological research, Resin application, Epoxy lay-up, furniture finishing
    Length (English) 10.5 in.
    Length (Metric) 26.7cm
    Material Vinyl, PVC
    Powdered Yes
    Size Small
    Thickness 10mil
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit