MAPA™ Solo Ultra 8 mil/980 Powder-Free Disposable Nitrile Gloves

Manufacturer: MAPA™ 980426

 View more versions of this product

Catalog No. 19-130-2652B

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Rolled
    Hand Ambidextrous
    For Use With (Application) handling food manufacturing medicine, splash protection in labs, assembly of small wet parts
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Nitrile
    Powdered No
    Size Small
    Thickness 8mil
    Coating Material Textured
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit