Learn More
MAPA™ TitanLite™ Gloves
Outlasts and outperforms traditional work gloves
Manufacturer:  MAPA™ 397427
 View more versions of this product
Catalog No. 19-049-138
Add to cart
Description
Description
For light- to medium-duty applications.
  • Comfortable interlock cotton liner has a light coating of nitrile for superior resistance to cuts, snags, punctures, and abrasion
  • Coated on the palm and fingers only, leaving the back of the glove uncoated for excellent ventilation and cool hands
  • Anatomically formed to the shape of the hand, reducing hand fatigue
  • Treated with Actifresh™ for resistance to bacteria, molds, and mildew. Launderable, with a smooth grip
  • With knitwrist
Compliance
Meet CE standards.
Specifications
Specifications
Safety
Knit
Nitrile
7
Palm, finger
light to medium duty applications
Cotton
Yellow