Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

MAPA™ Ultrane™ 560 Polyurethane Coated Gloves

Polyurethane coating provides maximum grip in oily or wet applications

Manufacturer:  MAPA™ 560391

 View more versions of this product

Catalog No. 19-249-735



 
Add to cart

Specifications Description & Specifications

Specifications

Entire hand
Launderable at 104°F with standard Detergent for reuse
Knit
Polyurethane
11
9.25 in.
Level 1
Smooth
handling Glass window panes, sheet Metal applications and any operation with a moderate risk of cuts and contact with Oily/wet parts
HDPE, Polyamide
Gray
23.49cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit