Learn More
MAPA™ Ultrane 551 Lightweight Polyurethane Coated Seamless Knit Gloves
Seamless knit lining absorbs perspiration for increased user comfort, while the polyurethane coating enhances grip capabilities.
Manufacturer:  MAPA™ 551400
 View more versions of this product
Catalog No. 19-161-408
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Reduced glove thickness increases dexterity, while still providing abrasion resistance
  • Seamless knit lining absorbs perspiration for increased user comfort
  • Silicone free to prevent traces, defects on metal sheets and glass prior to painting
  • Contains traces of DMF in compliance with occupational exposure limits
  • Free of harmful substances: STANDARD 100 by OEKO-TEX™
Specifications
Specifications
Gray
Ultrane 551
4131X
Specific
Seamless Knit
Automotive Assembly, Electronics Repair, and Construction
10
Polyurethane Foam
No Texture