Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK2 (ERK2) Mouse anti-Human, Rat, Mouse, Clone: 1B3B9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05157I25UL
Description
MAPK2 (ERK2) Monoclonal antibody specifically detects MAPK2 (ERK2) in Human, Rat, Mouse samples. It is validated for Immunoprecipitation, Western Blotting.Specifications
MAPK2 (ERK2) | |
Monoclonal | |
0.5 mg/mL | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
HPLC purified recombinant mouse p42MAP kinase (ERK2). | |
25 μL | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. | |
IgG2a κ |
Immunoprecipitation, Western Blot | |
1B3B9 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Human, Rat, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction