Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK2 (ERK2) Mouse anti-Human, Rat, Mouse, Clone: 1B3B9, MilliporeSigma™

Mouse Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen MAPK2 (ERK2)
Clone 1B3B9
Concentration 0.5 mg/mL
Applications Immunoprecipitation, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05157I100UL
View Documents
MilliporeSigma
05157I100UL
100 μL
Each for $373.00
Add to cart
 
05157I25UL
View Documents
MilliporeSigma
05157I25UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

MAPK2 (ERK2) Monoclonal antibody specifically detects MAPK2 (ERK2) in Human, Rat, Mouse samples. It is validated for Immunoprecipitation, Western Blotting.
Specifications

Specifications

MAPK2 (ERK2)
0.5 mg/mL
Unconjugated
Mouse
Human, Rat, Mouse
HPLC purified recombinant mouse p42MAP kinase (ERK2).
Monoclonal
Protein G purified
1B3B9
Immunoprecipitation, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit