Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mapk8ip2, Mouse anti-Mouse, PE/ATTO594, Clone: S135-37, Abnova™
Mouse Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18652
Specifications
Mapk8ip2 | |
S135-37 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
3230402N03Rik|AI847694|AW049958|Jip2|R75148 | |
Mapk8ip2 | |
Recombinant protein corresponding to amino acids 226-421 of mouse Mapk8ip2. | |
Monoclonal | |
100 μg | |
Store at -20°C. | |
Antibody |
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot | |
PE/ATTO594 | |
Liquid | |
mitogen-activated protein kinase 8 interacting protein 2 | |
60597 | |
Mouse | |
IgG1 | |
Primary | |
RUO | |
Human,Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title