Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mapk8ip2 Mouse anti-Human, Mouse, FITC, Clone:S135-37, Abnova™

Mouse monoclonal antibody raised against partial recombinant mouse Mapk8ip2.

Manufacturer:  Abnova Corporation MAB16884

Catalog No. 89-333-322


Add to cart

Specifications

Specifications

Mapk8ip2
S135-37
mitogen-activated protein kinase 8 interacting protein 2
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Liquid
Mouse
IgG1
Store at -20°C.
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
FITC
Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Mapk8ip2
3230402N03Rik, AI847694, AW049958, Jip2, R75148
Recombinant protein corresponding to amino acids 226-421 of mouse Mapk8ip2.
100μg
Antibody
60597
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit