Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPKAPK2 Antibody (7H4.2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $746.50
Specifications
Antigen | MAPKAPK2 |
---|---|
Clone | 7H4.2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:20 |
Applications | Immunoprecipitation, Western Blot |
Description
MAPKAPK2 Monoclonal specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MAPKAPK2 | |
1 mg/mL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
Human, Rat, Mouse | |
EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 | |
MAPKAPK2 | |
IgG | |
Affinity Purified | |
45 kDa |
7H4.2 | |
Western Blot 1:100-1:2000, Immunoprecipitation 1:20 | |
Monoclonal | |
Mouse | |
Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
P49137 | |
9261 | |
KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title