Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAPK2 Recombinant Human Protein Non-distribution product as customer accommodation.

MAPKAPK2 Recombinant Human Protein

$434.00

Specifications

Packaging 1 Each
Regulatory Status For research use only. Not for use in diagnostic procedures
Storage Requirements –80°C
Validated Application Kinase assay
Host Species Human
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3317 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3317
10μg Each for $434.00
Add to cart
 
Description

Description

MAPKAPK2 mediates p38 and ERK signaling in vivo. It is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation, and cell proliferation.
Specifications

Specifications

1 Each
–80°C
Human
For research use only. Not for use in diagnostic procedures
Kinase assay

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit