Learn More
MAPKAPK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154241AP
Catalog No. 50-173-0218
Add to cart
Description
Description
This gene encodes the general transcription factor IIB, one of the ubiquitous factors required for transcription initiation by RNA polymerase II. The protein localizes to the nucleus where it forms a complex (the DAB complex) with transcription factors IID and IIA. Transcription factor IIB serves as a bridge between IID, the factor which initially recognizes the promoter sequence, and RNA polymerase II.
Specifications
Specifications
MAPKAPK3
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16644, Q3UMW7, Q66H84
3PK, Chromosome 3p kinase, MAPKAP kinase 3, MAPKAP3, MAPKAPK 3, MAPKAPK3
Rabbit
IgG
150 μL
-20°C
Primary
102626, 315994, 7867
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
MAPKAPK3
Liquid
Mapkapk3
MAPKAPK3 Fusion Protein Ag7875
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat