Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPRE2 Monoclonal Antibody (KT52), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA547664

Encompass

Catalog No. PIMA547664


Add to Cart

Description

Description

The antibody detects a 36 kDa* protein corresponding to the molecular mass of EB2 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects EB2 by immunocytochemistry in HeLa and A431 cells.

MAPRE2 Monoclonal antibody specifically detects MAPRE2 in Human samples. It is validated for Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAPRE2
Monoclonal
Lot-Specific
PBS with 0.05% sodium azide
P23284
Mouse
Protein G
RUO
5479
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG2b
Immunocytochemistry, Western Blot
KT52
Unconjugated
PPIB
AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2
Clone M019 was generated from a proprietary antigen related to cyclophilin B from the MCF7 breast cancer cell line.
100 μL
Primary
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit