Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPRE2 Mouse anti-Canine, Human, Rat, Clone: OTI1A3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502796S

Catalog No. 50-167-3809


Add to cart

Description

Description

The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

MAPRE2
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15555, Q3B8Q0
CSCSC2, EB1, EB2, RP1
Mouse
IgG2b
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10982, 490487, 679221
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1A3
Unconjugated
MAPRE2
Liquid
Mapre2
Full length human recombit protein of human MAPRE2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit