Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marapsin/Pancreasin Rat anti-Mouse, Alexa Fluor™ 532, Clone: 15000000000, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221988AF532

Catalog No. NP221988A32


Add to cart

Description

Description

Marapsin/Pancreasin Monoclonal antibody specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for ELISA.
Specifications

Specifications

Marapsin/Pancreasin
15000000000
ELISA
Conjugated, Purified
PRSS27
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein Pancreasin. Rat monoclonal anti mouse Marapsin1 Q8BJR6.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27
Rat
IgG2
0.1 mL
Primary
83886.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit