Learn More
Marapsin/Pancreasin Rat anti-Mouse, Alexa Fluor™ 594, Clone: 15000000000, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221988AF594
Catalog No. NP221988A94
Add to cart
Description
Description
Marapsin/Pancreasin Monoclonal antibody specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for ELISA.
Specifications
Specifications
Marapsin/Pancreasin
15000000000
ELISA
CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
PRSS27
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein Pancreasin. Rat monoclonal anti mouse Marapsin1 Q8BJR6.
0.1 mL
Primary
83886