Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Marble Chips
SCI_ED

Catalog No. s25201a Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Solid
Suitable for use in Kipp Generator

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 472-34-1
Synonym Limestone, Natural Calcium Carbonate

Specifications

Quantity 500 g
Assay Percent Range >99%
Formula Weight 100.09
Packaging Solid
Density 2.83g/mL
Chemical Name or Material Marble Chips
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.