Learn More
Marburg Virus Mouse anti-Virus, DyLight 405, Clone: FM11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1107402V
Catalog No. NB1107402V
Add to cart
Description
Description
Marburg Virus Monoclonal specifically detects Marburg Virus in Virus samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Marburg Virus
FM11
Western Blot, ELISA
Purified
Mouse
IgG1
0.2 ml
Store at 4C in the dark.
Monoclonal
Virus
Western Blot, ELISA
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
MBG
Marburg virus.
Protein G purified
RUO
Primary
Marburg Virus (FM11)