Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCH9 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548850
Description
Predicted to react with mouse based on sequence homology.
E3 ubiquitin-protein ligase that may mediate ubiquitination of MHC-I, CD4 and ICAM1, and promote their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.Specifications
MARCH9 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q86YJ5 | |
MARCH9 | |
KLH conjugated synthetic peptide between 304-330 amino acids from the C-terminal region of human MARCH9. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MARCH9 | |
E3 ubiquitin-protein ligase MARCH9, 632-, Membrane-associated RING finger protein 9, Membrane-associated RING-CH protein IX, MARCH-IX, RING finger protein 179, MARCH9, RNF179 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92979 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction