Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCO Rabbit anti-Mouse, Alexa Fluor 350, Clone: 2359A, R&D Systems™

Rabbit Monoclonal Antibody

Manufacturer:  R&D Systems FAB29561U100UG

Catalog No. FAB29561U10


Add to cart
This item is not returnable. View return policy

Description

Description

MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

MARCO
2359A
Flow Cytometry 0.25-1 ug/106 cells
Q60754
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2
Mouse myeloma cell line, NS0-derived mouse MARCO protein, Gln70-Ser518, Accession # Q60754
Protein A or G purified from cell culture supernatant
RUO
Primary
8685
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Rabbit
IgG
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit