Learn More
Alfa Aesar™ Margatoxin, 99+%
Manufacturer:  Alfa Aesar™ J61103EXW
 View more versions of this product
Catalog No. AAJ61103EXW
Order in Bulk
Request bulk or custom formats
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
White
Powder
99+%
C178H286N52O50S7
rMargatoxin; MgTx
5 x 10ug
Peptide that blocks voltage-dependent Kv1.3 potassium channels
145808-47-5
4178.95
Margatoxin