Marigold Flock-Lined General-Purpose Latex Gloves
Liquidproof
Manufacturer: Protective Industrial Products, Inc. 48-326Y/XL
 View more versions of this product
Catalog No. 19-049276
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Latex
    Size X-Large
    Ideal for poultry processing; food processing; meat packing; fruit and vegetable growing; industrial plant maintenance; paper, optical, and pharmaceutical manufacturing; refinery and laboratory use; automotive and small parts assembly; electronics; and aircraft maintenance.
    • Beaded cuff for tear resistance and easy pull-on
    • Choice of yellow with diamond or pink with honeycomb palm pattern
    • Pink color is preferred by many workers in poultry and food processing
  • Description & Specifications
Related Products
Product Content Feedback