Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Marine Biology
An introduction to ocean ecosystems.
$15.00 - $17.00
Specifications
Category | Biology |
---|---|
Format | Book |
For Use With (Application) | Various ecosystem types are examined including coral reefs, estuaries, islands, tide pools and the abyss |
Description
- Vivid, full-color, detailed photographs and easy-to-understand explanations of marine biology
- Various ecosystem types are examined including coral reefs, estuaries, islands, tide pools, and the abyss
- 2nd Edition, 2002
- Student book: 144 pages
- Laboratory manual: 48 pages
- Teacher's guide: 56 pages
Specifications
Biology | |
Book | |
Various ecosystem types are examined including coral reefs, estuaries, islands, tide pools and the abyss |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title