Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte Marine Science Outfit
Learn about saline systems and brackish water.
$566.00
Specifications
For Use With (Application) | Designed for testing and studying saline systems (oceans, bays, salt marshes) and brackish water |
---|---|
Grade | 5+ |
Material | Fabric, Plastic |
Includes:
- Individual test modules for seven key salt water test factors
- Water sampling bottles
- Data sheets
- Three handbooks A Laboratory Manual for Marine Science, Limnology and Investigating Water Problems
- Soft-sided tote
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S85002
![]() |
LaMotte
590303 |
Marine science outfit |
Each for $566.00
|
|
Description
- Designed for testing and studying saline systems (oceans, bays, salt marshes) and brackish water
- Colorimetric test factor for pH (two ranges) and titration test factors for dissolved oxygen, hardness, carbon dioxide, alkalinity and salinity
- Soft-sided tote is made of durable fabric, solid plastic bottom ready for field work; handy side pockets hold thermometer, pens, writing tablets and other tools
Specifications
Designed for testing and studying saline systems (oceans, bays, salt marshes) and brackish water | |
5+ | |
Fabric, Plastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title