Learn More
MARK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154921AP
Catalog No. 50-173-0269
Add to cart
Description
Description
EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1. These isoforms were characterized by the presence of a 162-bp alternative exon that gave rise to two forms, one containing the exon and the other one lacking it. Both forms were found to be coexpressed in a number of selected cell lines and tissue samples. The human EMK1 was shown to be encoded by a single mRNA ubiquitously expressed.
Specifications
Specifications
MARK2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08679, Q05512, Q7KZI7
ELKL motif kinase 1, EMK 1, EMK1, MARK2, PAR 1, PAR1 homolog, Par1b
Rabbit
IgG
150 μL
-20°C
Primary
13728, 2011, 60328
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
MARK2
Liquid
Mark2
MARK2 Fusion Protein Ag7884
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat