Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ MARK3 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer: Abnova™ H00004140T02
Specifications
Western Blot | |
MARK3 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 81.4kDa | |
Transient overexpression cell lysate was tested with anti-MARK3 antibody (Mfr. No. H00004140-D01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
BC024773.1 | |
pCMV-MARK3 full-length | |
100 μL | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title