Learn More
MARK3 Mouse anti-Human, Clone: OTI3E11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805881
Catalog No. 50-166-7601
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is activated by phosphorylation and in turn is involved in the phosphorylation of tau proteins MAP2 and MAP4. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
MARK3
Monoclonal
Unconjugated
MARK3
Lyophilized
MARK3
Human recombit protein fragment corresponding to amino acids 473-729 of human MARK3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI3E11
PBS with 8% trehalose and no preservative; pH 7.3
P27448
CTAK1, KP78, PAR1A, Par-1a
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4140