Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mark3 Mouse anti-Human, Mouse, Rat, FITC, Clone: OTI2G8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272590F
Description
Mark3 Monoclonal specifically detects Mark3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
Mark3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MARK3 | |
Recombinant protein fragment corresponding to amino acids 473-729 of human Mark3 (NP_002367) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI2G8 | |
Immunohistochemistry | |
Cdc25C-associated protein kinase 1, cTAK1, C-TAK1, CTAK1ELKL motif kinase 2, EC 2.7.11, EC 2.7.11.1, EMK2, EMK-2, KP78, MAP/microtubule affinity-regulating kinase 3, PAR1A, Protein kinase STK10, Ser/Thr protein kinase PAR-1, Serine/threonine-protein kinase p78 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
4140.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title