Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARK4 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: OTI9B7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272591C
Description
MARK4 Monoclonal specifically detects MARK4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
MARK4 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MARK4 | |
Recombinant protein fragment corresponding to amino acids 390-467 of human MARK4 (NP_113605) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI9B7 | |
Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.1, FLJ12177, FLJ90097, KIAA1860MARK4L, MAP/microtubule affinity-regulating kinase 4, MAP/microtubule affinity-regulating kinase like 1, MAP/microtubule affinity-regulating kinase-like 1, MARK4 serine/threonine protein kinase, MARKL1L, MARKL1MARK4S, Nbla00650 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
57787.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title