Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517565
Description
It is not recommended to aliquot this antibody.
MARK4 is a serine/threonine kinase that is likely to be directly involved in microtubule organization in neuronal cells. The encoded protein is associated with the centrosome throughout mitosis and may be involved in cell cycle control. Expression of MARK4 is a potential marker for cancer, and the encoded protein may also play a role in Alzheimer's disease. Pseudogenes of MARK4 are located on both the short and long arm of chromosome 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for MARK4.
Specifications
MARK4 | |
Polyclonal | |
Unconjugated | |
Mark4 | |
2410090P21Rik; C79806; FLJ90097; KIAA1860; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; Mark4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 4; Nbla00650; PAR-1D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
232944, 57787, 680407 | |
-20°C | |
Liquid |
Western Blot | |
33 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q8CIP4, Q96L34 | |
Mark4 | |
Synthetic peptide corresponding to the sequence surrounding Cys514 of human MARK4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction