Learn More
Market Forge Market Forge Sterilmatic™ Sterilizer Accessory, Exhaust Condensers
For use with Sterilmatic Sterilizer
Manufacturer:  Market Forge 953629
 View more versions of this product
Catalog No. 14-460-262
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Requires : Drain, 13mm (1/2 in.) FPT or 16mm (5/8 in.) O.D.; copper steam exhaust connection, 10mm (3/8 in.) IPS
Description
Description
    • Condensing exhausted steam from sterilizer when unit cannot be vented to outside
    • Sterilizer can be used anywhere without having to worry about direct venting access
Certifications
UL listed, CSA certified
Specifications
Specifications
Sterilmatic Sterilizer
Exhaust Condenser
50Hz
Export
0-207kPa (0-30psi) pressure gauge, temperature gauge, low water cut-off, safety valve, thermostatic steam trap, signal light
220V