Learn More
Market Forge Market Forge Sterilmatic™ Sterilizer Accessory, Exhaust Condensers
For use with Sterilmatic Sterilizer
$2750.00
Specifications
Product Type Exhaust Condenser
For Use With (Equipment) Sterilmatic Sterilizer
View More Specs
Requires: Drain, 13mm (1/2 in.) FPT or 16mm (5/8 in.) O.D.; copper steam exhaust connection, 10mm (3/8 in.) IPS
Products 1
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
14-460-15
View Documents
Market Forge
95 0436
Domestic
Each for $2,750.00
Add to cart
Description
Description
    • Condensing exhausted steam from sterilizer when unit cannot be vented to outside
    • Sterilizer can be used anywhere without having to worry about direct venting access
Order Info
115V 50/60Hz
Specifications
Specifications
Exhaust Condenser
Sterilmatic Sterilizer