Learn More
Brady™ Marking Tape Roll: Abrasion Resistant Vinyl, Solid Color, Black
Ultra strong B-726 Abrasion-Resistant Vinyl tape makes your facility safer, more visual and more efficient by adding colorful visual indicators to aisles and passageways.
Manufacturer:  Brady™ 102839
Catalog No. 19-103-646
Add to cart
Description
Description
  • Pressure sensitive rubber based adhesive sticks tightly to virtually any clean, dry surface
  • Good chemical, oil and water resistance
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Quick application
Specifications
Specifications
No Text
Roll of 36 Yards
Vinyl
36 yd.
4 in.
Acrylic
Safety
Black
Marking Tape
0.004 in.
115.57 mm