Learn More
Brady™ Marking Tape Roll: Adhesive Vinyl, Solid Color, Black
Permanent acrylic adhesive sticks to a wide variety of surfaces
Manufacturer:  Brady™ 121131
Catalog No. 19-103-849
Add to cart
Description
Description
  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability
Specifications
Specifications
Roll of 50 Feet
ASTM D 2979
Black
Marking Tape
78.74 mm
Safety
No Text
Vinyl
50 ft.
Permanent Acrylic