Learn More
Brady™ Marking Tape Roll: Adhesive Vinyl, Solid Color, Blue
Permanent acrylic adhesive sticks to a wide variety of surfaces
Manufacturer:  Brady™ 121123
Catalog No. 19-103-841
Add to cart
Description
Description
  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability
Specifications
Specifications
Roll of 50 Feet
ASTM D 2979
Blue
Marking Tape
0.25 in.
Permanent Acrylic
Safety
No Text
Vinyl
50 ft.
6.4 mm