Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Marking Tape Roll: Adhesive Vinyl, Solid Color, Gray

Permanent acrylic adhesive sticks to a wide variety of surfaces

Supplier:  Brady™ 102830

Catalog No. 19-103-637

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability
Specifications

Specifications

Gray
3 in.
36 yd./Roll
Blank
Marking Tape
111.76 mm
0.004 in.
Vinyl
108 ft.
Acrylic
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit